missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TM 5275 sodium salt

Product Code. 18708219
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18708219 10 mg 10mg
18719927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18708219 Supplier Tocris Bioscience™ Supplier No. 5769/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Plasminogen activator inhibitor-1 (PAI-1) inhibitor; orally bioavailable

TM 5275 sodium salt is a plasminogen activator inhibitor-1 (PAI-1) inhibitor. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-[[2-[2-[4-(Diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt
CAS 1103926-82-4
Quantity 10 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C28H27ClN3NaO5
Formula Weight 543.97
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.