Learn More
TLX3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17454-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The TLX3 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TLX3. This antibody recognizes human, mouse, rat antigen. The TLX3 antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
TLX3 | |
Polyclonal | |
Unconjugated | |
20 μL | |
30012 | |
IgG |
Immunoprecipitation, ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |