missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TLX3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583146

Product Code. 16350105

  • £477.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: APFEDAGSYS VNLSLAPAGV IRVPAHRPLP GAVPPPLPSA LPAMPSVPTV SSLGGLNFPW MESSRRFVKD RFTA Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX3, also known as homeobox11-like1 (HOX11L2), is a DNA-binding nuclear transcription factor that is expressed in a subset of the primary sensory nervous system. TLX3, along with the related TLX1, is a selector transcription factor that promotes an excitatory glutamatergic neuronal phenotype over an inhibitory GABAergic phenotype opposing LBX1 signals during dorsal spinal cord development. Chromosomal translocations of the TLX3 gene have been shown to result in some forms of T-cell acute lymphoblastic leukemia (T-ALL).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TLX3
Polyclonal
Unconjugated
TLX3
homeo box 11-like 2; homeobox 11-like 2; homeobox protein Hox-11L2; Homeobox TLX-3; HOX11L2; Respiratory neuron homeobox protein; RGD1564190; RNX; T cell leukemia, homeobox 3; T-cell leukemia homeobox 3; T-cell leukemia homeobox protein 3; T-cell leukemia, homeobox 3; Tlx1l2; TLX3
Rabbit
Antigen affinity chromatography
RUO
30012
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O43711
TLX3
Recombinant protein corresponding to Human TLX3. Recombinant protein control fragment (Product #RP-108335).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.