missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559372
This item is not returnable.
View return policy
Description
Immunogen sequence: GIDQILSGPE TPGGGLGLGR GGQGHGENGA FSGGYHGASG YGPAGSL Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 74%.
The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX2, also known as homeobox11-like 1 (HOX11L1), is a DNA-binding nuclear transcription factor that appears to be crucial for the development of the peripheral nervous system. TLX2 is a downstream target of regulation by both PHOX2A and PHOX2B proteins during intestinal neuronal differentiation. In humans, loss-of-function of TLX2 may play a role in tumorigenesis of gastrointestinal stromal tumors.
Specifications
| TLX2 | |
| Polyclonal | |
| Unconjugated | |
| TLX2 | |
| Enteric neuron homeobox protein; Enx; homeo box 11-like 1; homeobox 11-like1; homeobox protein Hox-11L1; Homeobox TLX-2; hox11l; Hox11L.1; Hox11l1; NCX; Ncx1; neural crest homeobox protein; PMUR10F; T cell leukemia homeobox 2; T cell leukemia, homeobox 2; T-cell leukemia associated homeodomain protein; T-cell leukemia associated homeodomain protein Tlx3a; T-cell leukemia homeobox 2; T-cell leukemia homeobox protein 2; T-cell leukemia, homeobox 2; T-cell leukemia, homeobox 3a; Tlx1l1; Tlx1l2; Tlx2; tlx3a; Tlx-3a; tlxA; transcription factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3196 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43763 | |
| TLX2 | |
| Recombinant protein corresponding to Human TLX2. Recombinant protein control fragment (Product #RP-98392). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction