missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534553
This item is not returnable.
View return policy
Description
A suggested positive control is mouse spleen tissue lysate. PA5-34553 can be used with blocking peptide PEP-1596.
The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX1, also called homeobox11 (HOX11), as it is located outside of the four mammalian Hox clusters, is a DNA-binding nuclear transcription factor. TLX1 encodes a homeobox-domain containing protein and is containing a glycine and proline-rich amino terminus. TLX1 is required for maintenance of the developing spleen and cell survival. TLX1 is highly expressed in T-cell leukemia and lymphoid neoplasias as a result of translocation. TLX1-deficient mice have no spleen.
Specifications
| TLX1 | |
| Polyclonal | |
| Unconjugated | |
| TLX1 | |
| homeo box-11 (T-cell leukemia-3 associated breakpoint, homologous to Drosophila Notch); homeobox protein Hox11; homeobox protein Hox-11; homeobox TLX-1; HOX11; Hox-11; LOW QUALITY PROTEIN: T-cell leukemia homeobox protein 1; MGC163402; proto-oncogene TCL-3; RGD1563655; SI:dZ46P4.1; T cell leukemia homeobox 1; T cell leukemia, homeobox 1; T-cell leukemia homeobox 1; T-cell leukemia homeobox protein 1; T-cell leukemia, homeobox 1; T-cell leukemia/lymphoma protein 3; TCL3; TLX1; Tlx-1; transcription factor; ztlx-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21908, 3195, 499361 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P31314, P43345 | |
| TLX1 | |
| A 16 amino acid peptide near the amino terminus of human TLX1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction