missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TLX Recombinant Rabbit Monoclonal Antibody (24GB605)
GREENER_CHOICE

Product Code. 30276075
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30276075 100 μL 100µL
30276295 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30276075 Supplier Invitrogen™ Supplier No. MA557131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TLX Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLX
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB605
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NR2E1
Gene Accession No. Q64104, Q9Y466
Gene Alias fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991
Gene Symbols NR2E1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NR2E1 (aa 10-42).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21907, 684085, 7101
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.