Learn More
Invitrogen™ TLX Recombinant Rabbit Monoclonal Antibody (24GB605)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557132
 View more versions of this product
Product Code. 30276295
  • £126.00 / 20µL
Description
TLX Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLX
Recombinant Monoclonal
Unconjugated
NR2E1
fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991
Rabbit
Affinity Chromatography
RUO
21907, 684085, 7101
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB605
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q64104, Q9Y466
NR2E1
A synthesized peptide derived from human NR2E1 (aa 10-42).
20 μL
Primary
Human, Mouse, Rat