missing translation for 'onlineSavingsMsg'
Learn More

TLX/NR2E1 Rabbit anti-Human, Mouse, Rat, Clone: 6P3H8, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £386.00

Specifications

Antigen TLX/NR2E1
Clone 6P3H8
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18326173
View Documents
Bio-Techne
NBP3-16504-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18360634
View Documents
Novus Biologicals
NBP3-16504-100UL
100 μg
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TLX/NR2E1 Monoclonal antibody specifically detects TLX/NR2E1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

TLX/NR2E1
Western Blot
Unconjugated
Rabbit
Neuroscience
PBS, 0.05% BSA, 50% glycerol, pH7.3
7101
IgG
Affinity purified
6P3H8
Monoclonal
Purified
RUO
Human, Mouse, Rat
hTll, nuclear receptor subfamily 2, group E, member 1, Nuclear receptor TLX, Protein tailless homolog, tailes-related receptor, tailless homolog, Tll, TLXnuclear receptor subfamily 2 group E member 1, XTLL
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TLX/NR2E1 (Q9Y466).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.