missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9 Antibody (980914), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB36582MFV500
386.75 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TLR9 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 54106 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 980914 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Human TLR9 synthetic peptide, Accession # Q9NR96 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction