missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TLR8 Polyclonal antibody specifically detects TLR8 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | TLR8 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a peptide corresponding to 16 amino acids near the middle of human TLR8. The immunogen is located within amino acids 790 - 840 of TLR8. Amino Acid Squence: CASPGDQRGKSIVSLE |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?