missing translation for 'onlineSavingsMsg'
Learn More

TLR8 Antibody (44C143), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18411636 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411636 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411636 Supplier Novus Biologicals Supplier No. NBP224917MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR8 Monoclonal antibody specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry, Flow (Cell Surface), Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR8
Applications Flow Cytometry
Classification Monoclonal
Clone 44C143
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Flow (Cell Surface), Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
Host Species Mouse
Immunogen This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51311
Target Species Human, Mouse, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.