missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR8 Antibody (44C143), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24917AF647
This item is not returnable.
View return policy
Description
TLR8 Monoclonal specifically detects TLR8 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| TLR8 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Q9NR97 | |
| TLR8 | |
| This antibody was developed against a KLH-conjugated synthetic peptide of human TLR8, within amino acids 750-850. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 51311 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 44C143 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
| CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human, Mouse. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido