missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 (CD287) Mouse anti-Mouse, PE, Clone: A94B10, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565557
This item is not returnable.
View return policy
Description
TLR7 (CD287) Monoclonal antibody specifically detects TLR7 (CD287) in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
| TLR7 (CD287) | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| A94B10 | |
| PE | |
| TLR7; Tlr7 ; Toll-like receptor 7; CD287 | |
| Mouse TLR7 | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction