missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£158.00 - £302.00
Specifications
| Antigen | TLR7 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Flow Cytometry 1-2 ug/ 10^6 cells, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25586463) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18196681
|
Novus Biologicals
NBP2-24767SS |
0.025 mg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18186961
|
Novus Biologicals
NBP2-24767 |
0.1 mg |
£302.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR7 Polyclonal specifically detects TLR7 in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TLR7 | |
| Western Blot 0.5-2 ug/ml, Flow Cytometry 1-2 ug/ 10^6 cells, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25586463) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Primate, Rhesus Macaque | |
| toll-like receptor 7 | |
| TLR7 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and rhesus monkey, 83% homologous in dog and 72% homologous in mouse. |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 51284 | |
| A portion of amino acids 900-950 of human TLR7 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title