missing translation for 'onlineSavingsMsg'
Learn More

TLR7 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18410216 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410216 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410216 Supplier Novus Biologicals Supplier No. NBP224906MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TLR7 Polyclonal antibody specifically detects TLR7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR7
Applications Flow Cytometry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias toll-like receptor 7
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human TLR7 protein (between amino acids 700-750) [UniProt 9NYK1]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 51284
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.