missing translation for 'onlineSavingsMsg'
Learn More

TLR7 Antibody, Allophycocyanin, Novus Biologicals™

Product Code. 18430112
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430112 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430112 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224906APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TLR7 Polyclonal antibody specifically detects TLR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Proximity Ligation Assay, Dot Blot, Flow (Intracellular), Bioactivity
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Proximity Ligation Assay, Dot Blot, Flow Cytometry (Intracellular Staining), Bioactivity
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias toll-like receptor 7
Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 706-749 of human TLR7; GenBank no. AAF78035.1, AF245702_1. Cross-reactivity with mouse TLR7 shown. In human Ramos cells, additional bands are also seen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 51284
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.