missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody (6E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26284-50ul
This item is not returnable.
View return policy
Description
TLR7 Monoclonal antibody specifically detects TLR7 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| TLR7 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 6E5 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| toll-like receptor 7 | |
| A synthesized peptide derived from Human TLR7 [UniProt Q9NYK1] | |
| 50 μL | |
| Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| 51284 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction