missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody (4G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27332SS
147.05 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), CyTOF-ready, Knockdown Validated.
Specifications
| TLR7 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 10ug/ml∼, Flow Cytometry 1-2ug/10^6cells, Immunohistochemistry 1:50 - 1:200. Use reported in scientific literature (PMID 31092820), Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Flow (Intracellular) 5 ug/ml, CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 31730654) | |
| toll-like receptor 7 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. Mouse reactivity reported in scientific literature (PMID: 24126361). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF, KnockDown | |
| 4G6 | |
| Unconjugated | |
| Q9NYk1 | |
| TLR7 | |
| A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen. | |
| 0.025 mg | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| 51284 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction