missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody (4F4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04349CL1
This item is not returnable.
View return policy
Description
TLR7 Monoclonal antibody specifically detects TLR7 in Human samples. It is validated for Flow Cytometry,ELISA,Immunocytochemistry/Immunofluorescence
Specifications
| TLR7 | |
| Monoclonal | |
| CoraFluor 1 | |
| toll-like receptor 7 | |
| Recombinant human TLR7 (451-500aa) purified from E. coli | |
| 0.1 mL | |
| Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| 51284 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| 4F4 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction