missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLR6 Monoclonal Antibody (TLR6.127)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA140321
This item is not returnable.
View return policy
Description
MA1-40321 detects TLR6 from human samples. MA1-40321 has been successfully used in immunoprecipitation, immunofluorescence, and flow cytometry applications. The MA1-40321 immunogen is proprietary.
TLR6 (Toll-like receptor 6) is a member of the toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR6 functionally interacts with TLR2 to mediate cellular response to bacterial lipoproteins.
Specifications
| TLR6 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% BSA and no preservative | |
| Q9Y2C9 | |
| TLR6 | |
| Human Toll-like receptor 6. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| TLR6.127 | |
| Unconjugated | |
| TLR6 | |
| CD antigen 286; CD286; TLR6; toll like receptor 6; toll-like receptor 6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10333 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction