missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR6 Antibody (86B1153.2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56536AF488
This item is not returnable.
View return policy
Description
TLR6 Monoclonal specifically detects TLR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).
Specifications
| TLR6 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD286, CD286 antigen, toll-like receptor 6 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| TLR6 (86B1153.2) | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 86B1153.2 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular) | |
| Q9Y2C9 | |
| TLR6 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6. | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
| 10333 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction