missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR5 Antibody (624915), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6704CL1
This item is not returnable.
View return policy
Description
TLR5 Monoclonal antibody specifically detects TLR5 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
Specifications
| TLR5 | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
| E. coli-derived recombinant human TLR5, Ile21-Phe115, Accession # O60602 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Prostate Cancer | |
| 7100 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 624915 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction