missing translation for 'onlineSavingsMsg'
Learn More

TLR5 Antibody (19D759.2), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18415487 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415487 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415487 Supplier Novus Biologicals Supplier No. NBP224787MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR5 Monoclonal antibody specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Flow Cytometry, Flow (Cell Surface), Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR5
Applications Flow Cytometry
Classification Monoclonal
Clone 19D759.2
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Flow (Cell Surface), Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5
Host Species Mouse
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 between amino acids 700-800. The antibody also reacts with mouse TLR5.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7100
Target Species Human, Mouse, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.