missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56581PE
This item is not returnable.
View return policy
Description
TLR4 Polyclonal specifically detects TLR4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| TLR4 | |
| Polyclonal | |
| Flow Cytometry | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
| Rabbit | |
| 75 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7099 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| TLR4 | |
| This TLR4 antibody was developed against a synthetic peptide made to an internal portion of the mouse TLR4 protein (between residues 400-450) [Q9QUK6]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction