missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TLR4 Polyclonal specifically detects TLR4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | TLR4 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 |
| Gene Symbols | TLR4 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to the C-terminal portion of the human TLR4 protein (between residues 650-710) [Uniprot: O00206] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?