missing translation for 'onlineSavingsMsg'
Learn More

TLR3 Antibody (40C1285.6), PE, Novus Biologicals™

Product Code. 18400202 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400202 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400202 Supplier Novus Biologicals Supplier No. NBP224875PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLR3 Monoclonal antibody specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow (Intracellular), Bioactivity, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 40C1285.6
Conjugate PE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Bioactivity, Block/Neutralize
Formulation PBS
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 55-85 of human TLR3 was used as immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7098
Target Species Human, Mouse, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.