missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody (27N3D4) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-27404
This item is not returnable.
View return policy
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TLR3 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 2-4ug/ml, Flow Cytometry | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 7098 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Flow Cytometry | |
| 27N3D4 | |
| Unconjugated | |
| Q99MB1 | |
| TLR3 | |
| A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction