missing translation for 'onlineSavingsMsg'
Learn More

TLR3 Antibody (27N3D4), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18427545
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427545 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427545 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP227405AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

TLR3 Monoclonal antibody specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 27N3D4
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Host Species Rat
Immunogen A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7098
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.