missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody (002), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90455AF750
This item is not returnable.
View return policy
Description
TLR3 Monoclonal specifically detects TLR3 in Mouse samples. It is validated for ELISA.
Specifications
| TLR3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 2 | |
| ELISA | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TLR3 (NP_569054.2; Met1-Leu705). | |
| 0.1 mL | |
| Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Necroptosis, Signal Transduction | |
| 7098 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction