missing translation for 'onlineSavingsMsg'
Learn More

TLR2 Antibody (TL2.1), DyLight 680, Novus Biologicals™

Product Code. 18334323 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334323 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334323 Supplier Novus Biologicals Supplier No. NBP312062FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR2
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone TL2.1
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Host Species Rabbit
Immunogen This antibody was raised by immunising BALB/c mice with CHO cells transfected with human TLR2 cDNA.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7097
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.