missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (TL2.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 61 publication
Brand: Novus Biologicals NB100-56722
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional, In vitro assay, Dot Blot, Flow (Cell Surface), Block/Neutralize.
Specifications
| TLR2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 1 ug/10^6 cells, ELISA reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug, Functional reported in scientific literature (PMID 25412776), In vitro assay reported in scientific literature (PMID 24512642), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (PMID 24836676), Block/Neutralize reported in scientific literature (PMID 25412776) | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Functional Assay | |
| TL2.1 | |
| Unconjugated | |
| O60603 | |
| TLR2 | |
| This antibody was raised by immunizing mice with CHO cells transfected with human TLR2 cDNA (Flo et al, 2000). The hybridoma supernatants were selected by flow cytometry. | |
| 0.1 mg | |
| Immunology, Innate Immunity, Signal Transduction | |
| 7097 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction