missing translation for 'onlineSavingsMsg'
Learn More

TLR2 Antibody (TL2.1) - BSA Free, Novus Biologicals™

Product Code. 18054274 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054274 0.025 mg 25µg
18741684 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054274 Supplier Novus Biologicals Supplier No. NB10056722SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 61 publication

TLR2 Monoclonal specifically detects TLR2 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional, In vitro assay, Dot Blot, Flow (Cell Surface), Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR2
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone TL2.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Flow Cytometry 1 ug/10^6 cells, ELISA reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug, Functional reported in scientific literature (PMID 25412776), In vitro assay reported in scientific literature (PMID 24512642), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (PMID 24836676), Block/Neutralize reported in scientific literature (PMID 25412776)
Gene Accession No. O60603
Gene Alias CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Gene Symbols TLR2
Host Species Mouse
Immunogen This antibody was raised by immunizing mice with CHO cells transfected with human TLR2 cDNA (Flo et al, 2000). The hybridoma supernatants were selected by flow cytometry.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7097
Target Species Human, Mouse, Rat, Canine, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.