missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (67N8F8) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
£143.00 - £348.00
Specifications
| Antigen | TLR2 |
|---|---|
| Clone | 67N8F8 |
| Concentration | 0.5mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18140155
|
Novus Biologicals
NBP2-25259SS |
0.025 mg |
£143.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18123753
|
Novus Biologicals
NBP2-25259 |
0.1 mg |
£348.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| TLR2 | |
| 0.5mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| NP_003255.2 | |
| 7097 | |
| Amino acids 227-371 of human TLR2 were used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 67N8F8 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| TLR2 | |
| IgG1 κ | |
| Protein G purified | |
| This product reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title