missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (67N17F8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80980
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TLR2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot : 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin : 1:10-1:500 | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 67N17F8 | |
| Unconjugated | |
| PBS | |
| TLR2 | |
| Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody. | |
| 0.1 mg | |
| Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 7097 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction