missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLR11 Polyclonal Antibody

Description
A suggested positive control is RAW264.7 cell lysate. PA5-20057 can be used with blocking peptide PEP-0176.
The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules (1). The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition (2). TLR11 does not respond to known TLR ligands but instead respond specifically to uropathogenic bacteria. TLR11-deficient mice are highly susceptible to infection of the kidneys by uropathogenic bacteria, indicating that TLR11 may play a protective role in preventing infection of internal organs of the urogenital system.
Specifications
Specifications
| Antigen | TLR11 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Tlr11 |
| Gene Accession No. | Q6R5P0 |
| Gene Alias | Gm1365; Gm287; RGD1564460; RP23-392K10.4; Tlr11; Tlr12; Toll-like receptor 11; Toll-like receptor 12 |
| Gene Symbols | Tlr11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?