missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLR10 Monoclonal Antibody (670719)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561694
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is most highly expressed in lymphoid tissues such as spleen, lymph node, thymus, and tonsil. Its exact function is not known. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| TLR10 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9BXR5 | |
| TLR10 | |
| E. coli-derived recombinant human TLR10, Asn168-Lys383. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 670719 | |
| Unconjugated | |
| TLR10 | |
| CD290; MGC104967; MGC126398; MGC126399; TLR10; toll like receptor 10; toll-like receptor 10; UNQ315/PRO358 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 81793 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction