missing translation for 'onlineSavingsMsg'
Learn More

TLR1 Antibody (285923) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30734234 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734234 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734234 Supplier Novus Biologicals Supplier No. FAB1475MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

TLR1 Monoclonal antibody specifically detects TLR1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR1
Applications Western Blot
Classification Monoclonal
Clone 285923
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TLR1, Ser25-Asp581, Accession # Q9EPQ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7096
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.