missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TLR1 Monoclonal antibody specifically detects TLR1 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TLR1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 285923 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1 |
| Host Species | Rat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse TLR1, Ser25-Asp581, Accession # Q9EPQ1 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?