missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TLR (Cell Surface) Screening Antibody Pack

Product Code. 18174415 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
6 Vials
Unit Size:
Pack of 8
Product Code. Quantity unitSize
18174415 6 Vials Pack of 8
1 options
This item is not returnable. View return policy

Product Code. 18174415

Brand: Novus Biologicals™ NBP225086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • 1.NB100-56536: TLR6 Antibody (86B1153.2)
  • 2.NB100-56563: TLR1 Antibody
  • 3.NB100-56566: TLR4 Antibody (76B357.1)
  • 4.NB100-56722: TLR2 Antibody (TL2.1)
  • 5.NBP1-70343: TLR10 Antibody (3C10C5)
  • 6.NBP2-24787: TLR5 Antibody (19D759.2)

For Research applications

TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR (Cell Surface) Screening
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Flow (Cell Surface), Flow (Intracellular)
Immunogen TLR1: 25 ug: WB, Flow: H, M, R: Purified TLR2: 25 ug: WB, Flow, IP, IF/ICC: H, D: Purified TLR4: 25 ug: WB, Flow, IHC: H, M, R: Purified TLR5: 25 ug: WB, Flow, IHC: H, M, D: Purified TLR6: 25 ug: Flow, IHC: H: Purified TLR10: 25 ug: Flow: H: Purified WB:Western Blot IHC:Immunohistochemistry IP:Immunoprecipitation IF/ICC:Immunofluorescence/Immunocytochemistry D:Dog H:Human M:Mouse Mk:Monkey R:Rat
Quantity 6 Vials
Includes 1.NB100-56536: TLR6 Antibody (86B1153.2)
2.NB100-56563: TLR1 Antibody
3.NB100-56566: TLR4 Antibody (76B357.1)
4.NB100-56722: TLR2 Antibody (TL2.1)
5.NBP1-70343: TLR10 Antibody (3C10C5)
6.NBP2-24787: TLR5 Antibody (19D759.2)
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.