missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21546-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. Unlike other family members, a similar protein in mice does not cleave procollagen C-propeptides or chordin.Specifications
| TLL2 | |
| Polyclonal | |
| Unconjugated | |
| TLL2 | |
| KIAA0932, TLL2, tolloid like 2, Tolloid like protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7093 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y6L7 | |
| TLL2 | |
| TLL2 Fusion Protein Ag16209 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction