Learn More
Invitrogen™ Human TLK2, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human TLK2 (388-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for TLK2 is NP_006843.2.
Specifications
Specifications
| Accession Number | Q86UE8 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 11011 |
| Molecular Weight (g/mol) | 68.2 kDa |
| Name | Human TLK2, GST Tag |
| Quantity | 10 μg |
| Gene Alias | 4933403M19Rik; HsHPK; PKUalpha; PKU-alpha; protein kinase U-alpha; serine/threonine-protein kinase tousled-like 2; Tlk; Tlk2; tousled like kinase 2; tousled-like kinase 2; tousled-like kinase 2 (Arabidopsis) |
| Common Name | TLK2 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.