missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539444
This item is not returnable.
View return policy
Description
TLK2, a member of the Ser/Thr protein kinase family, is rapidly and transiently inhibited by phosphorylation following the generation of DNA double-stranded breaks during S-phase. This is cell cycle checkpoint and ATM-pathway dependent and appears to regulate processes involved in chromatin assembly. The TLK2 enzyme is cell-cycle regulated, with maximal activity in S-phase. It is inactivated by phosphorylation at Ser-750, potentially by CHK1. TLK2 heterodimerizes with TLK1. This nuclear protein is widely expressed, with presence in fetal placenta, liver, kidney, pancreas, heart and skeletal muscle tissues, and in several adult cell lines.
Specifications
| TLK2 | |
| Polyclonal | |
| Unconjugated | |
| TLK2 | |
| Serine/threonine-protein kinase tousled-like 2, Tousled-like kinase 2, PKU-alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11011, 24086 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86UE8, O55047 | |
| TLK2 | |
| A synthetic peptide derived from the internal region of human TLK2 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction