missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLK2 Monoclonal Antibody (OTI2G1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526736
This item is not returnable.
View return policy
Description
TLK2 Monoclonal Antibody for Western Blot, IHC (P)
TLK2 or Tousled-like kinase 2, is a nuclear serine/threonine kinases that is thought to be involved in the regulation of chromatin assembly. In addition, TLK2 is also involved in other cellular processes such as intracellular signaling cascade, protein amino acid phosphorylation, cell cycle, chromatin modification and in response to DNA damage stimulus. TLK2 displays maximal activity during S phase of the cell cycle where it appears to be regulated by cell-cycle-dependent phosphorylation. Inhibition of DNA replication causes a rapid loss of TLK2 activity, indicating that TLK2 function is tightly linked to ongoing DNA replication. The TLK2 enzyme is cell-cycle regulated, with maximal activity in S-phase. It is inactivated by phosphorylation at Ser-750, potentially by CHK1. TLK2 heterodimerizes with TLK1. This nuclear protein is widely expressed, with presence in fetal placenta, liver, kidney, pancreas, heart and skeletal muscle tissues, and in several adult cell lines.
Specifications
| TLK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86UE8 | |
| Tlk2 | |
| Human recombinant protein fragment corresponding to amino acids 1-296 of TLK2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| Tlk2 | |
| 4933403M19Rik; HsHPK; PKUalpha; PKU-alpha; protein kinase U-alpha; serine/threonine-protein kinase tousled-like 2; Tlk; Tlk2; tousled like kinase 2; tousled-like kinase 2; tousled-like kinase 2 (Arabidopsis) | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11011 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction