missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLK2 Antibody (OTI2D10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74532AF488
This item is not returnable.
View return policy
Description
TLK2 Monoclonal specifically detects TLK2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| TLK2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TLK2 | |
| Recombinant protein fragment corresponding to amino acids 1-296 of human TLK2 (NP_006843) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI2D10 | |
| Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.1, MGC44450, PKU-ALPHA, serine/threonine-protein kinase tousled-like 2, tousled-like kinase 2serine/threonine kinase | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 11011.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction