missing translation for 'onlineSavingsMsg'
Learn More

TLK117, MedChemExpress

Product Code. 16458457
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16458457

Brand: Medchem Express HY13634B5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TLK117, the active metabolite of TLK199, selective inhibits Glutathione S-transferase P1-1 (GSTP1-1) with a Ki of 0.4 μM for GSTP. TLK117 also competitively inhibits glyoxalase I with a Ki of 0.56 μM.

TLK117, the active metabolite of TLK199, selective inhibits Glutathione S-transferase P1-1 (GSTP1-1) with a Ki of 0.4 μM for GSTP. TLK117 also competitively inhibits glyoxalase I with a Ki of 0.56 μM.

Specifications

Chemical Name or Material TLK117
Physical Form Solid
CAS 152684-53-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H27N3O6S
For Use With (Application) Cancer-programmed cell death
Synonym TER117
Solubility Information DMSO : 150 mg/mL (316.76 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](C1=CC=CC=C1)NC([C@@H](NC(CC[C@H](N)C(O)=O)=O)CSCC2=CC=CC=C2)=O
Molecular Weight (g/mol) 473.54
Formula Weight 473.54
Percent Purity 98.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.