missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | TLK1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 μL/mg of lysate |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617595
|
Novus Biologicals
NBP2-98826-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648295
|
Novus Biologicals
NBP2-98826-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLK1 Polyclonal antibody specifically detects TLK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| TLK1 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Repair, Tumor Suppressors | |
| 0.2 um filtered solution in PBS | |
| 9874 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.1, KIAA0137serine/threonine-protein kinase tousled-like 1, PKU-beta, SNARE protein kinase SNAK, tousled-like kinase 1serine threonine protein kinase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human TLK1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title