missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35294-100ul
This item is not returnable.
View return policy
Description
TLE4 Polyclonal antibody specifically detects TLE4 in Rat samples. It is validated for ELISA,Western Blot
Specifications
| TLE4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| BCE1, BCE-1, E(spI), ESG4, homolog of Drosophila E(sp1), KIAA1261, Protein BCE-1, transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila), transducin-like enhancer protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 173-243 of human TLE4 (NP_001269677).,, Sequence:, SSALGGQSHLPIKDEKKHHDNDHQRDRDSIKSSSVSPSASFRGAEKHRNSADYSSESKKQKTEEKEIAARY | |
| 100 μL | |
| Signal Transduction | |
| 7091 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction