missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22094-1-AP-20UL
This item is not returnable.
View return policy
Description
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.Specifications
| TLE3 | |
| Polyclonal | |
| Unconjugated | |
| TLE3 | |
| ESG, ESG3, GRG3, HsT18976, KIAA1547, TLE3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21887, 7090, 84424 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q04726, Q08122, Q9JIT3 | |
| TLE3 | |
| TLE3 Fusion Protein Ag17300 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction