missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLE3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540528
This item is not returnable.
View return policy
Description
Peptide sequence: GSSSGLLALG ALGSQAHLTV KDEKNHHELD HRERESSANN SVSPSESLRA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%; Zebrafish: 79%.
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.
Specifications
| TLE3 | |
| Polyclonal | |
| Unconjugated | |
| TLE3 | |
| 2610103N05Rik; enhancer of split groucho 3; enhancer of split groucho-like protein 3; ESG; ESG-1; ESG-2; ESG3; ESG-3; ESG-4; Esp3; GRG3; Grg-3; Grg3a; Grg3b; HsT18976; KIAA1547; mKIAA1547; rTLE3; TLE family member 3, transcriptional corepressor; Tle1; Tle2; Tle3; tle4; transducin like enhancer of split 3; transducin-like enhancer of split 3; transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3 homolog of Drosophila; transducin-like enhancer of split 3 short isoform; transducin-like enhancer of split 3, E(spl) homolog; transducin-like enhancer of split 3, homolog of Drosophila E(sp1); transducin-like enhancer of split 3, homolog of Drosophila E(spl); transducin-like enhancer protein 3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7090 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q04726 | |
| TLE3 | |
| Synthetic peptide directed towards the N-terminal of human TLE3 (aa 161-210). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction