missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE3 Mouse anti-Human, Clone: 1D11A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66083-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.Specifications
TLE3 | |
Monoclonal | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04726 | |
TLE3 | |
TLE3 Fusion Protein Ag17906 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1D11A8 | |
Unconjugated | |
TLE3 | |
ESG, ESG3, GRG3, HsT18976, KIAA1547, TLE3 | |
Mouse | |
Protein G | |
RUO | |
7090 | |
-20°C | |
Liquid |